missing translation for 'onlineSavingsMsg'
Learn More

anti-IL-8 Oligoclonal, Invitrogen™

IL-8Oligoclonal antibody 710371 from Thermo Scientific™ Pierce™ specifically detects IL-8 in Porcine samples, and it is validated for WB

Marca:  Invitrogen 710371

Dettagli aggiuntivi : Netto-Gewicht : 0.01000kg

Codice prodotto. 15327324

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Descrizione

Descrizione

Interleukin 8 is a pro-inflammatory factor which belongs to the CXC subfamily of chemokinesk, and which are bound by the cell surface receptors IL-8RA and IL-8RB. IL-8 functions as a chemoattractant and is a potent angiogenic factor. The expression and secretion of IL-8 can be induced by diverse inflammatory stimuli in many cells, including macrophages and endothelial cells. In endothelial cells, IL-8 is present in storage vesicles called Weibel-Palade bodies.

710371 detects IL-8 in Porcine samples. 710371 has been successfully used in WB procedures. The immunogen for 710371 is recombinant protein corresponding to amino acids 26-103 of swine IL-8.

Applications:

Western Blotting

Specifica

Specifica

IL-8
19HCLC
P26894
Recombinant protein corresponding to amino acids 26-103 of swine IL-8
Primary
Porcine
Purified
Polyclonal
Unconjugated
Rabbit
Protein A
396880
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali