Learn More
IL-35 Recombinant Rabbit Monoclonal Antibody (6H15L18), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 701101
Weitere Details : Netto-Gewicht : 0.00450kg
Beschreibung
Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
IL-35 Recombinant Monoclonal antibody specifically detects IL-35 in Human samples. It is validated for Western Blot.Spezifikation
IL-35 | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P29459, Q14213 | |
EBI3, IL12A | |
Recombinant protein corresponding to amino acids 57–186 of human interleukin 35 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
6H15L18 | |
Unconjugated | |
EBI3 | |
CLMF; CLMF p35; cytokine receptor; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; EBI3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; epstein-Barr virus-induced gene 3 protein; IL-12 subunit p35; IL-12, subunit p35; IL12A; IL-12A; IL12p35; IL27 subunit; IL-27 subunit beta; IL27B; IL-27B; il35; IL35 subunit; ILN; Interleukin; interleukin 12, p35; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; interleukin-12 subunit alpha; Interleukin-27 subunit beta; Interleukin-35; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35 | |
Rabbit | |
Protein A | |
RUO | |
10148, 3592 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |