missing translation for 'onlineSavingsMsg'
Learn More

IL-12 p35 Recombinant Polyclonal Antibody (2H6L2), Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Marke:  Invitrogen 710154

Weitere Details : Netto-Gewicht : 0.00450kg

Artikelnummer. 15307224

  • 469.00 € / 100 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

This antibody is predicted to react with human, mouse and rat based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

IL-12 p35 Recombinant Polyclonal antibody specifically detects IL-12 p35 in Non-human Primate samples. It is validated for Western Blot.
Spezifikation

Spezifikation

IL-12 p35
Recombinant Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide
P29459, P48091
IL12A
Recombinant protein corresponding to amino acids 23–328 of monkey IL-12
100 μg
Primary
Non-human Primate
Antibody
IgG
Western Blot
2H6L2
Unconjugated
IL12A
CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35
Rabbit
Protein A
RUO
3592, 703205
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote