Learn More
IL17RD Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547287
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse (rm) IL-17 RD is observed and less than 10% cross-reactivity with recombinant human (rh) IL-17 RC, rmIL-17B R, and rhIL-17 R is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
IL17RD Polyclonal antibody specifically detects IL17RD in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IL17RD | |
Polyclonal | |
Unconjugated | |
Il17rd | |
2810004A10Rik; AI428510; HH18; hSef; IL-17 receptor D; IL17RD; IL-17RD; IL17Rhom; IL17RLM; interleukin 17 receptor D; interleukin-17 receptor D; Interleukin-17 receptor-like protein; mSef; RGD1565237; SEF; sef homolog; Sef-S; UNQ6115/PRO20026 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
54756 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8NFM7 | |
Il17rd | |
Mouse myeloma cell line NS0-derived recombinant human IL-17RD/SEF Ala27-Arg299 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |