Learn More
IL-1RAcP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528051
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2 membrane extract, mouse liver. Predicted reactivity: Mouse (88%), Rat (88%), Rhesus Monkey (99%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IL-1RAcP Polyclonal antibody specifically detects IL-1RAcP in Human, Mouse, Porcine samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
IL-1RAcP | |
Polyclonal | |
Unconjugated | |
IL1RAP | |
6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1R-3; IL1RAcP; IL-1RAcp; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100628318, 16180, 3556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q61730, Q9NPH3 | |
IL1RAP | |
Recombinant fragment corresponding to a region within amino acids 1 and 266 of Human IL1RAP | |
100 μL | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG |