Learn More
IL-34 Monoclonal Antibody (1D12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517098
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17098 targets IL34 in indirect ELISA, FACS, ICC, IF, and WB applications and shows reactivity with Human samples. The MA5-17098 immunogen is purified recombinant fragment of human IL34 expressed in E. Coli. MA5-17098 detects IL34 which has a predicted molecular weight of approximately 27.5kDa.
IL-34 Monoclonal antibody specifically detects IL-34 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
IL-34 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q6ZMJ4 | |
Il34 | |
Purified recombinant fragment of human IL34 expressed in E. Coli. | |
RUO | |
146433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
1D12 | |
Unconjugated | |
Il34 | |
2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |