missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL5RA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527900
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IL5RA Polyclonal antibody specifically detects IL5RA in Human samples. It is validated for Western Blot.Spezifikation
IL5RA | |
Polyclonal | |
Unconjugated | |
IL5RA | |
CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3568 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q01344 | |
IL5RA | |
Recombinant fragment corresponding to a region within amino acids 1 and 339 of Human IL5 Receptor alpha | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |