missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528135
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, H1299, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ILF1 Polyclonal antibody specifically detects ILF1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
ILF1 | |
Polyclonal | |
Unconjugated | |
Foxk2 | |
1110054H05Rik; 5730434B08Rik; 6230415M23Rik; cellular transcription factor ILF-1; forkhead box K2; forkhead box protein K2; FOXK1; foxk2; G/T-mismatch specific binding protein; ILF; ILF1; ILF-1; interleukin enhancer binding factor 1; interleukin enhancer-binding factor 1; nGTBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3607 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q01167 | |
Foxk2 | |
Synthetic peptide corresponding to a region within amino acids 598 and 660 of Human ILF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |