missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imipramine Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA175479
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-75479 specifically detects Imipramine. PA1-75479 has been successfully used in ELISA applications. The PA1-75479 immunogen is Imipramine-BTG.
Imipramine Polyclonal antibody specifically detects Imipramine in Chemical samples. It is validated for ELISA.Spezifikation
Imipramine | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Sheep | |
purified | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
Unconjugated | |
Antideprin; C19H24N2; Deprenil; Deprimin; Deprinol; Depsonil; Dynaprin; Eupramin; Imipramil; Irmin; Janimine; Melipramin; Surplix; Tofranil | |
Imipramine-BTG | |
1 mg | |
Primary | |
Antibody | |
IgG |