missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A NP Monoclonal Antibody (AA5H), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA123054
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-23054 detects Influenza A Virus Nucleoprotein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Influenza A NP Monoclonal antibody specifically detects Influenza A NP in Virus samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Influenza A NP | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
AA5H | |
Unconjugated | |
Influenza nucleoprotein | |
Influenza A / Puerto Rico /8/34 (H1N1) and A/Bangkok /1/79 (H3N2) viruses. | |
500 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |