missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H3N2 HA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534930
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Recommended positive controls: H3N2 HA recombinant protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Influenza A H3N2 HA Polyclonal antibody specifically detects Influenza A H3N2 HA in Virus samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Influenza A H3N2 HA | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; H3N2 HA; H3N2 hemagglutinin; Influenza H3N2; influenza hemagglutinin | |
Recombinant fragment corresponding to N-terminal of Influenza A virus HA (A/TW/3446/02(H3N2)) | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.64 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |