Learn More
Influenza B Virions Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA173036
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The PA1-73036 antibody reacts with Influenza B Virions. PA1-73036 has been successfully used in Immunofluorescence applications. The PA1-73036 antibody was raised against the Yamagata strain of Influenza B.
Influenza B Virions Polyclonal antibody specifically detects Influenza B Virions in Virus samples. It is validated for Immunocytochemistry.Spezifikation
Influenza B Virions | |
Polyclonal | |
FITC | |
Influenza Virions | |
Yamagata strain of Influenza B. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |