missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518054
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine based on sequence homology.
ING2 Polyclonal antibody specifically detects ING2 in Human samples. It is validated for Western Blot.Spezifikation
ING2 | |
Polyclonal | |
Unconjugated | |
Ing2 | |
2810011M06Rik; ING1L; ING1Lp; ING2; Ing2b; Inhibitor of growth 1-like protein; inhibitor of growth family member 2; inhibitor of growth family, member 2; inhibitor of growth protein 2; p32; p33ING2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3622 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H160 | |
Ing2 | |
Synthetic peptide sequence (DKSTEKTKKDRRSR) corresponding to the C-terminus amino acids of ING2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |