Learn More
ING3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540703
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LSSGTGAGAI TMAAAQAVQA TAQMKEGRRT SSLKASYEAF KNNDFQLGKE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 75%; Zebrafish: 92%
ING3 Polyclonal antibody specifically detects ING3 in Human samples. It is validated for Western Blot.Spezifikation
ING3 | |
Polyclonal | |
Unconjugated | |
ING3 | |
1300013A07Rik; Eaf 4; Eaf4; FLJ20089; HSPC301; ING 2; ING 3; ING2; ING3; ing3 protein; inhibitor of growth family member 3; inhibitor of growth family, member 3; inhibitor of growth protein 3; inhibitor of growth protein 3-like protein; MEAF4; p47 p47 ING3; p47 regulator protein; p47ING3; p47ING3 protein; zgc:56327 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54556 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NXR8 | |
ING3 | |
Synthetic peptide directed towards the middle region of human ING3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |