Learn More
ING3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527717
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: H1299, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ING3 Polyclonal antibody specifically detects ING3 in Human samples. It is validated for Western Blot.Spezifikation
ING3 | |
Polyclonal | |
Unconjugated | |
ING3 | |
1300013A07Rik; Eaf 4; Eaf4; FLJ20089; HSPC301; ING 2; ING 3; ING2; ING3; ing3 protein; inhibitor of growth family member 3; inhibitor of growth family, member 3; inhibitor of growth protein 3; inhibitor of growth protein 3-like protein; MEAF4; p47 p47 ING3; p47 regulator protein; p47ING3; p47ING3 protein; zgc:56327 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
Q9NXR8 | |
ING3 | |
Synthetic peptide corresponding to a region within amino acids 84 and 179 of Human ING3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |