Learn More
INPP1 Monoclonal Antibody (1A6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA519210
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: MSDILRELLC VSEKAANIAR ACRQQEALFQ LLIEEKKEGE KNKKFAVDFK TLADVLVQEV IKQNMENKFP GLEKNIFGEE SNEFTNDWGE KITLRLCSTE EETAELLSKV LNGNKVASEA LARVVHQDVA FTDPTLDSTE INVPQDILGI WVDPIDSTYQ YIKGSADIKS NQGIFPCGLQ CVTILIGVYD IQTGVPLMGV INQPFVSRDP NTLRWKGQCY WGLSYMGTNM HSLQLTISRR NGSETHTGNT GSEAAFSPSF SAVISTSEKE TIKAALSRVC GDRIFGAAGA GYKSLCVVQG LVDIYIFSED TTFKWDSCAA HAILRAMGGG IVDLKECLER NPETGLDLPQ LVYHVENEGA AGVDRWANKG GLIAYRSRKR LETFLSLLVQ NLAPAETHT
INPP1 Monoclonal antibody specifically detects INPP1 in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Spezifikation
INPP1 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P49441 | |
INPP1 | |
INPP1 (AAH15496, 1 a.a. ∽ 399 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation, Western Blot | |
1A6 | |
Unconjugated | |
INPP1 | |
2300002C06Rik; AV137389; Inositol polyphosphate 1-phosphatase; inositol polyphosphate-1-phosphatase; Inpp1; IPP; IPPase | |
Mouse | |
Affinity chromatography | |
RUO | |
3628 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |