Learn More
INPP4A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530072
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (92%), Rat (94%), Xenopus laevis (89%), Pig (97%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
INPP4A Polyclonal antibody specifically detects INPP4A in Human samples. It is validated for Western Blot.Spezifikation
INPP4A | |
Polyclonal | |
Unconjugated | |
INPP4A | |
107kDa; 4-Ptase-1; 9630012D15; D130048C09Rik; Inositol polyphosphate 4-phosphatase type I; Inositol polyphosphate 4-phosphatase-1; Inositol polyphosphate-4-phosphatase type I A; inositol polyphosphate-4-phosphatase, type 1; inositol polyphosphate-4-phosphatase, type I; inositol polyphosphate-4-phosphatase, type I, 107kD; inositol polyphosphate-4-phosphatase, type I, 107kDa; INPP4; Inpp4a; R74740; TVAS1; Type I inositol 3,4-bisphosphate 4-phosphatase; type I inositol-3; type I inositol-3,4-bisphosphate 4-phosphatase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3631 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96PE3 | |
INPP4A | |
Recombinant protein fragment corresponding to a region within amino acids 304 and 489 of Human INPP4A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |