missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSRR Monoclonal Antibody (326929), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524051
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
INSRR Monoclonal antibody specifically detects INSRR in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
INSRR | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P14616 | |
Insrr | |
Mouse myeloma cell line NS0-derived recombinant human INSRR Leu24-Leu923 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
326929 | |
Unconjugated | |
Insrr | |
Insrr; insulin receptor related receptor; insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IRR; IR-related receptor; Sirr | |
Mouse | |
Protein A/G | |
RUO | |
3645 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |