Learn More
IDE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529350
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, H1299, HeLa, HepG2, mouse brain, rat brain. Predicted reactivity: Mouse (94%), Rat (95%), Zebrafish (90%), Dog (93%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IDE Polyclonal antibody specifically detects IDE in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IDE | |
Polyclonal | |
Unconjugated | |
IDE | |
1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15925, 25700, 3416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.6 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P14735, P35559, Q9JHR7 | |
IDE | |
Recombinant fragment corresponding to a region within amino acids 349 and 635 of Human insulin degrading enzyme | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |