missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin/Proinsulin Monoclonal Antibody (D3E7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA116710
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Insulin/Proinsulin Monoclonal antibody specifically detects Insulin/Proinsulin in Bovine, Human, Porcine, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen).Spezifikation
Insulin/Proinsulin | |
Monoclonal | |
4.9 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01308, P01315, P01317, P01322, P01323 | |
INS, Ins1, Ins2 | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human insulin. | |
200 μg | |
Primary | |
Bovine, Human, Porcine, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Frozen) | |
D3E7 | |
Unconjugated | |
INS | |
AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin | |
Mouse | |
Protein A | |
RUO | |
24505, 24506, 280829, 3630, 397415 | |
4° C, do not freeze | |
Liquid |