Learn More
INSR alpha Monoclonal Antibody (83-14), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA513756
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-13756 targets Insulin Receptor alpha in WB, ICC/IF, ELISA and IP applications and shows reactivity with Human samples. The MA5-13756 immunogen is iM-9 lymphocytes followed by purified insulin receptor.
INSR alpha Monoclonal antibody specifically detects INSR alpha in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation.Spezifikation
INSR alpha | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P06213 | |
INSR | |
IM-9 lymphocytes followed by purified insulin receptor | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Immunoprecipitation | |
83-14 | |
Biotin | |
INSR | |
4932439J01Rik; CD220; CD220 beta; D630014A15Rik; DAF-2; HHF5; INSR; insulin receptor; insulin receptor (INSR); insulin receptor precursor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR alpha; IR beta; ir-A; IR-B | |
Mouse | |
Protein A | |
RUO | |
3643 | |
4° C | |
Liquid |