Learn More
INSR alpha Mouse anti-Bovine, Human, Ovine, Porcine, Rabbit, Rat, Biotin, Clone: 83-7, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA513764
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-13764 targets Insulin Receptor alpha in ELISA, FACS, ICC/IF and IHC (P) applications and shows reactivity with Bovine, Human, Ovine, Porcine, Rat and Rabbit samples. The MA5-13764 immunogen is iM-9 lymphocytes followed by IR.
The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a beta-alpha-alpha-beta configuration. The beta-subunit (95kDa) possesses a single transmembrane domain, whereas the alpha-subunit (135kDa) is completely extracellular.Spezifikation
INSR alpha | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P06213, P15127 | |
INSR | |
IM-9 lymphocytes followed by IR | |
500 μL | |
Primary | |
Bovine, Human, Ovine, Porcine, Rabbit, Rat | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
83-7 | |
Biotin | |
INSR | |
CD220, HHF5, IR, IR alpha | |
Mouse | |
Protein G | |
RUO | |
100008911, 24954, 3643, 396755, 408017, 443431 | |
4° C | |
Liquid |