missing translation for 'onlineSavingsMsg'
Learn More

anti-Integrin alpha-2b/CD41 APC, Clone: eBioMWReg30 (MWReg30), Invitrogen™

Integrin alpha-2b/CD41 Monoclonal antibody A16229 from Invitrogen specifically detects Integrin alpha-2b/CD41 in Mouse samples. It is validated for Flow Cytometry.

Marke:  Invitrogen A16229

Weitere Details : Netto-Gewicht : 0.00280kg

Artikelnummer. 15245984

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »


Beschreibung

Beschreibung

Integrin alpha-2b/CD41 Monoclonal antibody A16229 from Invitrogen specifically detects Integrin alpha-2b/CD41 in Mouse samples. It is validated for Flow Cytometry.

ITGA2B encodes integrin alpha chain 2b. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. Alpha chain 2b undergoes post-translational cleavage to yield disulfide-linked light and heavy chains that join with beta 3 to form a fibronectin receptor expressed in platelets that plays a crucial role in coagulation. Mutations that interfere with this role result in thrombasthenia. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG1, κ
Clone: eBioMWReg30 (MWReg30)
Immunogen: Mouse CD41 protein.
Formulation: 25μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: BDPLT2, CD41, CD41B, GP2B, GPIIb, GT, GTA, HPA3, GPalpha IIb, integrin alpha-IIb, platelet fibrinogen receptor, alpha subunit, platelet membrane glycoprotein IIb, platelet-specific antigen BAK, ITGA2B
Uniprot Accession: Q9QUM0
Entrez GeneID: 16399
Gene Symbol: Itga2b
This product detects Integrin alpha-2b/CD41 in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Mouse CD41 protein.
Spezifikation

Spezifikation

Integrin alpha-2b/CD41
Monoclonal
APC
Q9QUM0
Itga2b
Mouse CD41 protein.
Primary
Mouse
Purified
Flow Cytometry
eBioMWReg30 (MWReg30)
25μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
BDPLT2, CD41, CD41B, GP2B, GPIIb, GT, GTA, HPA3, GPalpha IIb, integrin alpha-IIb, platelet fibrinogen receptor, alpha subunit, platelet membrane glycoprotein IIb, platelet-specific antigen BAK, ITGA2B
Rat
25 μg
16399
Store at 4° C
IgG1 κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote