missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537435
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 25 and 129 kDa. Purity is >95% by SDS-PAGE.
ITGA7 Polyclonal antibody specifically detects ITGA7 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
ITGA7 | |
Polyclonal | |
Unconjugated | |
Itga7 | |
[a]7; alpha 7A integrin; alpha7; H36-alpha7; integrin alpha 7; integrin alpha 7 chain; integrin alpha-7; Integrin alpha-7 70 kDa form; Integrin alpha-7 heavy chain; Integrin alpha-7 light chain; integrin subunit alpha 7; integrin, alpha 7; ITGA7; UNQ406/PRO768 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16404, 3679, 81008 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q13683, Q61738, Q63258 | |
Itga7 | |
Synthetic peptide corresponding to amino acids 926-976 of human Integrin alpha7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |