Learn More
ITGA9/ITGB1 Monoclonal Antibody (Y9A2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA180920
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human Integrin alpha 9 beta 1 antibody, clone Y9A2 recognizes the human alpha9 integrin, a approximately 140 kDa glycoprotein expressed as a heterodimer in association with the beta1 integrin.
ITGA9/ITGB1 Monoclonal antibody specifically detects ITGA9/ITGB1 in Guinea Pig, Human, Porcine samples. It is validated for Flow Cytometry, Immunoprecipitation.Spezifikation
ITGA9/ITGB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P05556, Q13797, Q9GLP0 | |
Itga9, Itgb1 | |
alpha9 integrin transfected lambda cells. | |
200 μg | |
Primary | |
Guinea Pig, Human, Porcine | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
Y9A2 | |
Unconjugated | |
Itga9 | |
ALPHA-RLC; CD29; fibronectin receptor subunit beta; FNRB; glycoprotein IIa; GPIIA; integrin alpha 9; integrin alpha-9; Integrin alpha-RLC; integrin beta 1; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin subunit alpha 9; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin, alpha 9; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITGA4L; ITGA9; ITGB1; MDF2; MSK12; RLC; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
Protein A | |
RUO | |
100626707, 100716659, 100719502, 3680, 3688, 397019 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |