missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 Monoclonal Antibody (2G5.1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA121434
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD103 Monoclonal antibody specifically detects CD103 in Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Spezifikation
CD103 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA and 0.09% sodium azide | |
P38570 | |
Itgae | |
Human intestinal lymphocyte (HML1) | |
100 μg | |
Primary | |
Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
2G5.1 | |
Unconjugated | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Mouse | |
Protein A | |
RUO | |
102128361, 3682, 721483 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |