missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD51 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547096
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In Western blots, less than 1% cross-reactivity with recombinant human Integrin alpha 5 and recombinant mouse Integrin E is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD51 Polyclonal antibody specifically detects CD51 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
CD51 | |
Polyclonal | |
Unconjugated | |
ITGAV | |
1110004F14Rik; 2610028E01Rik; alpha v integrin; antigen identified by monoclonal antibody L230; Cd51; CD61; D430040G12Rik; Dnmt3l-ps1 pseudogene; GP3A; GPIIIa; integrin alpha V; integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; integrin subunit alpha V; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); ITGAV; ITGB3; MSK8; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); vitronectin receptor subunit alpha; VNR; VNRA; VTNR | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16410, 296456, 3685 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P06756, P43406 | |
ITGAV | |
CHO-derived recombinant human Integrin alpha V/CD51 Phe31-Val992 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |