Learn More
anti-Integrin alpha X/CD11c Alexa Fluor™ 488, Clone: N418, Invitrogen™
Integrin alpha X/CD11c Monoclonal antibody MCD11c20 from Invitrogen specifically detects Integrin alpha X/CD11c in Mouse samples. It is validated for Flow Cytometry.
Marke: Invitrogen MCD11C20
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Integrin alpha X/CD11c Monoclonal antibody MCD11c20 from Invitrogen specifically detects Integrin alpha X/CD11c in Mouse samples. It is validated for Flow Cytometry.
ITGAX is the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles.Host Species: Hamster
Cross Reactivity: Mouse
Isotype: IgG
Clone: N418
Immunogen: Mouse CD11c.
Formulation: 1mL Purified Antibody in PBS, with 0.1% sodium azide
Synonyms: AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Uniprot Accession: Q9QXH4
Entrez GeneID: 16411
Gene Symbol: Itgax
This product detects Integrin alpha X/CD11c in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Mouse CD11c.
Spezifikation
Integrin alpha X/CD11c | |
Monoclonal | |
Alexa Fluor 488 | |
Q9QXH4 | |
Itgax | |
Mouse CD11c. | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
N418 | |
1mL Purified Antibody in PBS, with 0.1% sodium azide | |
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X | |
Hamster | |
1 mL | |
16411 | |
Store at 4° C | |
IgG |