missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD61 Monoclonal Antibody (CRC54), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA135264
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 1 mL of distilled water.
CD61 Monoclonal antibody specifically detects CD61 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Spezifikation
CD61 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide | |
P05106 | |
ITGB3 | |
CD61. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Frozen) | |
CRC54 | |
Unconjugated | |
ITGB3 | |
antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
Mouse | |
purified | |
RUO | |
3690 | |
4° C | |
Lyophilized |