missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA184928
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
In Western blot, this antibody detects a band at ∽19kDa. Rabbit anti Chicken interferon alpha antibody recognizes chicken Interferon alpha (chIFNα).
IFN alpha Polyclonal antibody specifically detects IFN alpha in Chicken samples. It is validated for ELISA, Functional Assay, Immunoprecipitation, Western Blot.Spezifikation
IFN alpha | |
Polyclonal | |
Unconjugated | |
IFNA3 | |
alpha-2a interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN alpha 1/13; IFN alpha 1b; IFN a; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNa; INFA2; interferon; interferon alpha; interferon alpha 1; Interferon alpha 13 (IFNA13); interferon alpha 15; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; Interferon alpha leukocyte (IFL); interferon alpha protein; interferon alpha-1; Interferon alpha1 (IFNA1); interferon alpha-1 precursor; interferon alpha-1/13; Interferon alpha-1/13 (IFN-alpha-1/13); Interferon alpha-2; Interferon alpha-A; Interferon alpha-D; Interferon alpha-D (IFN alphaD); interferon type A1/A2; interferon type A3; interferon, alpha 1; | |
Rabbit | |
Protein A | |
RUO | |
396398 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Functional Assay, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with no preservative | |
Q90872 | |
IFNA3 | |
Recombinant chicken Interferon alpha | |
500 μg | |
Primary | |
Chicken | |
Antibody | |
IgG |