Learn More
CXCL10 Monoclonal Antibody (0D10), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen AHC2369
Weitere Details : Netto-Gewicht : 0.50000kg
Artikelnummer. 10261533
  • 454.00 € / 1 Stück
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Beschreibung
Beschreibung
This product is recommended as a detection antibody in Sandwich ELISA applications. This clone can also be referred to as clone number 96/2 OD10.
CXCL10 Monoclonal antibody specifically detects CXCL10 in Human samples. It is validated for ELISA.
Spezifikation
Spezifikation
CXCL10
Monoclonal
0.5 mg/mL
PBS with 5mg/mL BSA and 0.5mg/mL sodium azide; pH 7.4
P02778
CXCL10
Human recombinant IP-10.
100 μg
Primary
Human
Antibody
IgG1
ELISA
0D10
Biotin
CXCL10
10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C X C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; CRG-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamm
Mouse
Protein A
RUO
3627
4° C