Learn More
IP3 Receptor 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1904
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-904 detects inositol 1,4,5-trisphosphate receptor type-II (IP3R-II) from rat and mouse tissues. PA1-904 has been successfully used in immunohistochemistry, immunofluorescence and immunocytochemistry procedures. Immunohistochemical staining of IP3R-II in rat brain with PA1-904 results in staining of the hippocampus, corpus collosum, and cerebellum. This product fails to detect IP3R-II in Western blot procedures. PA1-904 immunizing peptide corresponds to amino acid residues 317-334 from rat IP3R-II. PA1-904 immunizing peptide (Cat. # PEP-024) is available for use in neutralization and control experiments.
IP3 Receptor 2 Polyclonal antibody specifically detects IP3 Receptor 2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen).Spezifikation
IP3 Receptor 2 | |
Polyclonal | |
Unconjugated | |
ITPR2 | |
AI649341; ANHD; CFAP48; cilia and flagella associated protein 48; inositol 1,4,5-triphosphate receptor 2; inositol 1,4,5-triphosphate receptor 5; inositol 1,4,5-triphosphate receptor, type 2; inositol 1,4,5-trisphosphate receptor type 2; inositol 1,4,5-trisphosphate receptor, type 2; inositol 1,4,5-trisphosphate type V receptor; inositol triphosphate receptor type 2; inositol trisphosphate receptor type 2; insP3R2; InsP3R-2; InsP3R-5; IP3 receptor; IP3 receptor isoform 2; IP3 receptor/Ca2+ channel type 2; IP3R; IP3R 2; IP3R2; Itpr2; Itpr5; LOW QUALITY PROTEIN: inositol 1,4,5-trisphosphate receptor type 2; si:dkey-196d8.1; Type 2 inositol 1,4,5-trisphosphate receptor; type 2 InsP3 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16439, 3709, 81678 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen) | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P29995, Q14571, Q9Z329 | |
ITPR2 | |
Synthetic Peptide: E(317) L N P D Y R D A Q N E G K T V R D(334). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |