Learn More
IPP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529510
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, U87-MG, SK-N-SH. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (80%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IPP Polyclonal antibody specifically detects IPP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IPP | |
Polyclonal | |
Unconjugated | |
IPP | |
Actin-binding protein IPP; IAP promoted placental; intracisternal A particle-promoted polypeptide; IPP; kelch-like family member 27; kelch-like protein 27; KLHL27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3652 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y573 | |
IPP | |
Recombinant fragment corresponding to a region within amino acids 122 and 406 of Human IPP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |