Learn More
INSR Recombinant Rabbit Monoclonal Antibody (6H23L15), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 701049
Weitere Details : Netto-Gewicht : 0.00450kg
Beschreibung
This antibody is predicted to react with mouse based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
INSR Recombinant Monoclonal antibody specifically detects INSR in Human samples. It is validated for Western Blot.Spezifikation
INSR | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B | |
Recombinant protein corresponding to amino acids 1283–1382 of human insulin receptor | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
6H23L15 | |
Unconjugated | |
P06213 | |
Rabbit | |
Protein A | |
RUO | |
3643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |