missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK-M Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517473
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
IRAK-M Polyclonal antibody specifically detects IRAK-M in Human, Non-human Primate samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
IRAK-M | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q9Y616 | |
IRAK3 | |
Synthetic peptide corresponding to residues near the carboxy-terminus of human IRAK-M | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
IRAK3 | |
4833428C18Rik; AI563835; ASRT5; FLJ13601; IL-1 receptor-associated kinase M; interleukin 1 receptor associated kinase 3; interleukin-1 receptor-associated kinase 3; IRAK3; IRAK-3; IRAKM; IRAK-M | |
Rabbit | |
Protein A | |
RUO | |
11213 | |
-20°C | |
Liquid |