missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA19057
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
IRAK4 Polyclonal antibody specifically detects IRAK4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.Spezifikation
IRAK4 | |
Polyclonal | |
Unconjugated | |
IRAK4 | |
8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
51135 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NWZ3 | |
IRAK4 | |
Synthetic peptide corresponding to the N terminus amino acids NKPITPSTYVRC. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |