Learn More
IRE1 alpha Polyclonal Antibody, DyLight™ 488, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA522726
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 13298818
  • 473.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
This antibody does not react with non-human primate samples. Suggested positive control: Pancreatic islets and insulinoma cells.
IRE1 alpha Polyclonal antibody specifically detects IRE1 alpha in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation
Spezifikation
IRE1 alpha
Polyclonal
DyLight 488
ERN1
9030414B18Rik; AI225830; C85377; endoplasmic reticulum (ER) to nucleus signalling 1; endoplasmic reticulum to nucleus signaling 1; endoplasmic reticulum to nucleus signalling 1; endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; FLJ30999; hIRE1p; inositol-requiring 1; inositol-requiring 1 alpha; inositol-requiring enzyme 1; inositol-requiring protein 1; IRE1; IRE1a; IRE1a antibody; Ire1alpha; Ire1-alpha; Ire1p; IRE1p antibody; MGC163277; MGC163279; protein kinase/endoribonuclease; RGD1559716; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1; serine/threonine-protein kinase/endoribonuclease IRE1-like
Rabbit
Antigen affinity chromatography
RUO
2081, 498013, 78943
4° C, store in dark
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
50mM sodium borate with no preservative
O75460, Q9EQY0
ERN1
A synthetic peptide within the human IRE1 alpha protein (within residues 700-800).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers