missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517252
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
IRF2 Polyclonal antibody specifically detects IRF2 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
IRF2 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P14316, P23906 | |
IRF2 | |
Synthetic peptide corresponding to residues surrounding Asp327 of human IRF-2 protein | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
IRF2 | |
9830146E22Rik; AI646973; DKFZp686F0244; Interferon regulatory factor 2; IRF2; Irf-2; OTTHUMP00000218576; OTTHUMP00000218580 | |
Rabbit | |
Affinity chromatography | |
RUO | |
16363, 290749, 3660 | |
-20°C | |
Liquid |