missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517580
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other family members at physiological levels.
IRF4 Polyclonal antibody specifically detects IRF4 in Human samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.Spezifikation
IRF4 | |
Polyclonal | |
Unconjugated | |
IRF4 | |
AI385587; interferon regulatory factor 4; Irf4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; multiple myeloma oncogene 1; MUM1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; SHEP8; Spip; Transcriptional activator PIP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3662 | |
-20°C | |
Liquid |
ChIP Assay, Immunoprecipitation, Western Blot | |
120 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q15306 | |
IRF4 | |
Synthetic peptide corresponding to residues around Asp175 of human IRF-4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |