missing translation for 'onlineSavingsMsg'
Learn More

IRF7 Recombinant Rabbit Monoclonal Antibody (1H31L43), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marke:  Invitrogen 700229

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15316954

  • 464.00 € / 100 Mikrogramm
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

IRF7 Recombinant Monoclonal antibody specifically detects IRF7 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

IRF7
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
IMD39; Interferon regulatory factor 7; interferon regulatory factor-7H; Irf7; IRF-7; IRF7A; IRF7B; IRF7C; IRF7H; IRF-7H
A recombinant protein corresponding to amino acids 269-378.
100 μg
Primary
Human
Liquid
Western Blot
1H31L43
Unconjugated
Q92985
Rabbit
Protein A
RUO
3665
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote