Learn More
IRF2BP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544309
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LVARNGEAEV SPTAGAEAVS GGGSGTGATP GAPLCCTLCR ERLEDTHFVQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
IRF2BP1 Polyclonal antibody specifically detects IRF2BP1 in Human samples. It is validated for Western Blot.Spezifikation
IRF2BP1 | |
Polyclonal | |
Unconjugated | |
Irf2bp1 | |
6330414O09Rik; C530033F24; DKFZP434M154; interferon regulatory factor 2 binding protein 1; interferon regulatory factor 2-binding protein 1; IRF-2-binding protein 1; IRF2BP1; IRF-2BP1; Probable E3 ubiquitin-protein ligase IRF2BP1; Probable RING-type E3 ubiquitin transferase IRF2BP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26145 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IU81 | |
Irf2bp1 | |
Synthetic peptide directed towards the middle region of human IRF2BP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |