missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRGM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA520507
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is rat brain tissue lysate. PA5-20507 can be used with blocking peptide PEP-0627.
IRGM Polyclonal antibody specifically detects IRGM in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
IRGM | |
Polyclonal | |
Unconjugated | |
IRGM | |
Ifggd3; Ifi1; Iigp3; Iipg3; immunity related GTPase M; immunity related GTPase m1; immunity-related GTPase family M member 1; immunity-related GTPase family M protein; Immunity-related GTPase family M protein 1; immunity-related GTPase family, M; immunity-related GTPase family, M1; immunity-related GTPase M; interferon inducible protein 1; interferon-gamma-inducible GTPase Ifggd3 protein; interferon-inducible GTPase 3; Interferon-inducible protein 1; Irgm; Irgm1; LPS-stimulated RAW 264.7 macrophage protein 47; LPS-stimulated RAW 264.7 macrophage protein 47 homolog; LRG 47; LRG47; LRG-47; LRG-47-like protein; MGC149263 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15944, 303090, 345611 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
A1A4Y4, Q60766, Q6AYC2 | |
IRGM, Irgm1 | |
A 17 amino acid peptide near the amino terminus of the human IRGM. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |