Learn More
WNT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534321
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Baboon, Monkey, Marmoset (100%).
WNT2 Polyclonal antibody specifically detects WNT2 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
WNT2 | |
Polyclonal | |
Unconjugated | |
Wnt2 | |
2610510E18Rik; etID309852.18; INT1L 1; Int1l1; int-1-like protein 1; INT-1-related protein; IRP; IRP protein; Mirp; protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; wingless-type MMTV integration site family, member 2; Wnt; WNT 2; Wnt family member 2; Wnt2; Wnt-2; Wnt2a; ZfWnt2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7472 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P09544 | |
Wnt2 | |
Synthetic 15 amino acid peptide from internal region of human WNT2. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |