missing translation for 'onlineSavingsMsg'
Learn More

ISG15 Monoclonal Antibody (T.991.5), Invitrogen™

Rabbit Monoclonal Antibody

Marke:  Invitrogen MA515029

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11505001

  • 472.00 € / 100 Mikroliter
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other ubiquitin family members, including ubiquitin, SUMO-1, SUMO-2, SUMO-3 and NEDD8.

ISG15 Monoclonal antibody specifically detects ISG15 in Human samples. It is validated for Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

ISG15
Monoclonal
129 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P05161
ISG15
Synthetic peptide corresponding to amino acids at the amino terminus of human ISG15 protein
100 μL
Primary
Human
Antibody
IgG
Immunoprecipitation, Western Blot
T.991.5
Unconjugated
ISG15
G1P2; hUCRP; IFI15; IGI15; IMD38; interferon, alpha-inducible protein; interferon, alpha-inducible protein (clone IFI-15K); interferon-induced 15 kDa protein; interferon-induced 15-KDa protein; interferon-induced 17 kDa protein; interferon-induced 17-kDa/15-kDa protein; interferon-stimulated protein (15 kDa); interferon-stimulated protein 15; interferon-stimulated protein, 15 kDa; IP17; Irfp; Isg15; ISG15 ubiquitin like modifier; ISG15 ubiquitin-like modifier; ubiquitin cross-reactive protein; Ubiquitin-like protein ISG15; UCRP
Rabbit
Affinity chromatography
RUO
9636
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote