missing translation for 'onlineSavingsMsg'
Learn More

IRF9 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA124390

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11540983

  • 492.00 € / 50 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1-24390 detects ISGF3G in Human samples. PA1-24390 has been successfully used in Western Blot procedures. PA1-24390 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human ISGF3G. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

ISGF3G functions to recruit RNA polymerase II to the promoter of interferon-stimulated genes and requires histone deacetylases. Defects in ISGF3 can cause resistance to IFN-alpha;(2a) treatment.
Spezifikation

Spezifikation

IRF9
Polyclonal
Unconjugated
Q00978
IRF9
Synthetic peptide region of human ISGF3G.
50 μg
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
1 mg/mL
IRF9
IRF-9, ISGF3, ISGF3G, p48
Rabbit
Antigen affinity chromatography
RUO
10379
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote