Learn More
ISL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541670
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MDSPCQPQPL SQALPQLPGS SSEPLEPEPG RARMGVESYL PCPLLPSYHC Sequence homology: Cow: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 85%
ISL2 Polyclonal antibody specifically detects ISL2 in Human samples. It is validated for Western Blot.Spezifikation
ISL2 | |
Polyclonal | |
Unconjugated | |
ISL2 | |
3110001N10Rik; Insulin gene enhancer protein ISL-2; insulin related protein 2 (islet 2); ISL LIM homeobox 2; ISL2; Isl-2; ISL2 transcription factor, LIM/homeodomain, (islet-2); islet 2; islet-2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64843 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96A47 | |
ISL2 | |
Synthetic peptide directed towards the N-terminal of human ISL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |