Learn More
ISL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527789
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: K562 whole cell extract, K562 nuclear extract, jurkat whole cell extract, jurkat nuclear extract, PC-12. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Japanese Medaka (97%), Xenopus laevis (94%), Chicken (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ISL1 Polyclonal antibody specifically detects ISL1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ISL1 | |
Polyclonal | |
Unconjugated | |
ISL1 | |
Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16392, 3670, 64444 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.55 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P61371, P61372, P61374 | |
ISL1 | |
Recombinant protein fragment corresponding to a region within amino acids 23 and 331 of Human Islet 1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |