missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoxsuprine Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA175223
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-75223 specifically detects Isoxsuprine. PA1-75223 has been successfully used in ELISA applications. The PA1-75223 immunogen is Isoxsuprine(p-OH)-BTG.
Isoxsuprine Polyclonal antibody specifically detects Isoxsuprine in Chemical samples. It is validated for ELISA.Spezifikation
Isoxsuprine | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Sheep | |
purified | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
Unconjugated | |
C18H23NO3; Dilavase; Duvadilan; Vasodilian; Vasosurpine | |
Isoxsuprine(p-OH)-BTG | |
1 mg | |
Primary | |
Antibody | |
IgG |