missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543052
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ILKRPARRSD MDRPEGPGEE GPGEAAASGS GLEKPPKDQP QEGRKSKRRV Sequence homology: Human: 100%; Rabbit: 83%; Rat: 77%
ISX Polyclonal antibody specifically detects ISX in Human samples. It is validated for Western Blot.Spezifikation
ISX | |
Polyclonal | |
Unconjugated | |
ISX | |
9130012O13Rik; DKFZp781N2395; intestine specific homeobox; intestine-specific homeobox; ISX; MGC138417; pancreas-intestine homeodomain transcription factor; Pix-1; RAX-like homeobox; RAXLX | |
Rabbit | |
Affinity Chromatography | |
RUO | |
91464 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2M1V0 | |
ISX | |
Synthetic peptide directed towards the N-terminal of human ISX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |