Learn More
CXCL11 Monoclonal Antibody (10C6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA1032
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
MA1032 has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody MA1033B (biotinylated conjugate of clone 22F1). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125-0.25 μg/mL for detection. The Molecular weight is ∽8303 Da from positions 22 - 94 as determined by MALDI.
CXCL11 Monoclonal antibody specifically detects CXCL11 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
CXCL11 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
O14625 | |
CXCL11 | |
Full length recombinant protein expressed in E-Coli | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
10C6 | |
Unconjugated | |
CXCL11 | |
betaR1; beta-R1; b-R1; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 11; CXC; CXC motif chemokine; C-X-C motif chemokine 11; C-X-C motif chemokine ligand 11; Cxc11; CXCL; CXCL11; H174; Interferon gamma-inducible protein 9; interferon-inducible T-cell alpha chemoattractant; IP9; IP-9; Itac; I-TAC; MGC102770; Scyb11; Scyb9b; small inducible cytokine B11; small inducible cytokine subfamily B (Cys-X-Cys), member 11; small inducible cytokine subfamily B (Cys-X-Cys), member 9B; small-inducible cytokine B11 | |
Mouse | |
Protein G | |
RUO | |
6373 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |